missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals

£25.63

Chemical Identifiers

CAS 78922-04-0
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acidShow MoreShow Less
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15439009
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14285.30
250 g
£25.63
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.