missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-410(Cl), ion exchange resin, Thermo Scientific Chemicals

£20.26 - £201.00

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423328
SDS View Documents
Thermo Scientific Alfa Aesar
A17734.22
100 g
£20.26
100g
Please to purchase this item. Need a web account? Register with us today!
11433328
SDS View Documents
Thermo Scientific Alfa Aesar
A17734.36
500 g
£52.16
500g
Please to purchase this item. Need a web account? Register with us today!
11443328
SDS View Documents
Thermo Scientific Alfa Aesar
A17734.0E
2500 g
£201.00
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9002-26-0
MFCD00132710

Specifications

9002-26-0
MFCD00132710
Amberlite IRA-410(Cl), ion exchange resin
Amine-like
14,382
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.