Learn More
Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Styrene-DVB; Mix of strongly acidic and basic gel type resin
£47.94 - £585.92
Specifications
Odor | Odorless |
---|---|
Molecular Formula | Styrene-DVB |
MDL Number | MFCD00145822 |
Solubility Information | Insoluble in water,acids and bases. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375327
|
Thermo Scientific Alfa Aesar
040992.36 |
500 g |
£47.94
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355327
|
Thermo Scientific Alfa Aesar
040992.A3 |
2 kg |
£136.68
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365327
|
Thermo Scientific Alfa Aesar
040992.A9 |
10 kg |
£585.92
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odorless | |
Styrene-DVB | |
MFCD00145822 | |
Insoluble in water,acids and bases. |
Safety and Handling
P280-P305+P351+P338-P310
H318
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only