missing translation for 'onlineSavingsMsg'
Learn More
Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals Product Code.: 11379379

Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals

Product Code. 11379379
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11379379

Brand: Thermo Scientific Alfa Aesar 089079.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strongly acidic macroreticular resin

Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39389-20-3
Molecular Formula C18H18O3S
Molecular Weight (g/mol) 314.399
MDL Number MFCD00145841
InChI Key SIWVGXQOXWGJCI-UHFFFAOYSA-N
Synonym amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
PubChem CID 170197
IUPAC Name 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
SMILES C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O

Specifications

Physical Form Pale brown beads
Odor Odorless
Color Brown
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ 15(H)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY22K045 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number089079.18,089079.A1,089079.30
Lot NumberZ11J072 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number089079.18,089079.A1,089079.30
Lot NumberX09G054 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number089079.18,089079.A1,089079.30
Lot NumberS22I043 Certificate TypeCertificate of Analysis Date20/07/2022 Catalog Number089079.18,089079.A1,089079.30
Lot NumberY30H006 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number089079.18,089079.A1,089079.30
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.