missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals

£38.20 - £499.00

Chemical Identifiers

CAS 39389-20-3
Molecular Formula C18H18O3S
Molecular Weight (g/mol) 314.399
MDL Number MFCD00145841
InChI Key SIWVGXQOXWGJCI-UHFFFAOYSA-N
Synonym amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
PubChem CID 170197
IUPAC Name 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
SMILES C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11345168
View Documents
Thermo Scientific Alfa Aesar
044079.30
250 g
£38.20
250g
Please to purchase this item. Need a web account? Register with us today!
11335168
View Documents
Thermo Scientific Alfa Aesar
044079.A1
1 kg
£119.00
1kg
Please to purchase this item. Need a web account? Register with us today!
11355168
View Documents
Thermo Scientific Alfa Aesar
044079.A7
5 kg
£499.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39389-20-3
314.399
SIWVGXQOXWGJCI-UHFFFAOYSA-N
170197
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
C18H18O3S
MFCD00145841
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid

Specifications

Beads
C18H18O3S
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
SIWVGXQOXWGJCI-UHFFFAOYSA-N
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
170197
39389-20-3
MFCD00145841
Insoluble in water.
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
314.399
Amberlyst™ 15 (H) ion exchange resin, Wet, Styrene-DVB; Strongly acidic macroreticular resin
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only