missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ HPR900, OH-form ion-exchange resin

Product Code. p-2885567 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10713151

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 197809-12-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197809-12-4
MDL Number MFCD00802695

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.45, harmonic mean size: 0.560 to 0.700mm,% <0.300mm: <0.5%,%>1.180mm: <5.0%
Packaging Plastic bottle
Volume Total Capacity volume capacity as is (OH):> 0.9 eq/L
Water Retention Capacity (OH): 66.0 to 75.0%
Color Ivory
Quantity 100 g
Chemical Name or Material Ambersep™ 900

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.