Learn More
Invitrogen™ Ambion™ 0.5-10kb RNA Ladder
Suitable for sizing single-stranded RNA ranging from 0.5kb to 10kb in glyoxal or formaldehyde agarose gels
Brand: Invitrogen™ 15623200
Code : 50
Additional Details : CAS Number : 79-06-1 Weight : 0.00454kg
Includes: 0.5-10kb RNA RNA Ladder is supplied at 1μg/μL in 10mM HEPES (pH 7.2), 2mM EDTA
Description
- Consists of nine bands in even increments (0.5, 1, 1.5, 2, 3, 4, 6, 8, and 10kb) to simplify size estimation
- Compared to other bands, 1.5kb band is more intense, allowing easy orientation and identification of RNA bands on gel
- RNA molecules in ladder are polyA-tailed (40 nt) for convenient use as control templates for cDNA synthesis
- Compatible with formaldehyde-containing, glyoxal-containing and polyacrylamide gels
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Dry Ice
Specifications
75 μL |
For Research Use Only. Not for use in diagnostic procedures.