Learn More
Invitrogen™ TBE (10X), RNase-free
Molecular biology grade, Invitrogen 10X TBE solution is supplied in four bottles containing 1 L each. The buffer is certified RNase-free, economical, and ready-to-use.
Brand: Invitrogen™ AM9863
74.24 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Molecular biology grade, Invitrogen 10X TBE solution is supplied in four bottles containing 1 L each. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Certifications
Certified RNase-free
Specifications
Running Buffer | |
Ambion | |
Molecular Biology |
10X | |
1 L |
Safety and Handling
- TBE (10X), RNase-free
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Boric acid
For Research Use Only. Not for use in diagnostic procedures.