Learn More
Invitrogen™ Barrier (Filter) Tips, 10 μL size (compatible with Pipetman™ pipettors)
Barrier tips prevent cross-contamination of samples by means of an aerosol filter.
Brand: Invitrogen™ AM12640
208.10 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : CAS Number : 9048-46-8 Weight : 1.02000kg
Includes: Ten racks containing 8 x 12 tips each
Description
- Tips are RNase- and DNase-free and ideal for handling RNA, PCR applications and working with radioisotopes or toxic chemicals
- Compatible with Pipetman™ pipetters
- Each tip lot is subjected to rigorous testing for RNase contamination and certified to be nuclease-free
Specifications
10 μL | |
Sterile | |
Barrier | |
Room Temperature | |
Invitrogen |
Filtered | |
Rack | |
Store at room temperature. | |
Gilson Pipetman |
For Research Use Only. Not for use in diagnostic procedures.