missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BSA, acetylated (20 mg/mL)

Product Code. 10117054
Click to view available options
Quantity:
6 x 25 mg
Unit Size:
150mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10117054

Brand: Invitrogen™ AM2614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Invitrogen Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminants such as nucleases and proteases is essential.

Invitrogen Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminants such as nucleases and proteases is essential. Six tubes of 25mg are provided, at a concentration of 40 to 50mg/mL. The acetylation process inactivates any trace nuclease activity that might be present. While the acetylation process does change BSA's binding characteristics and makes it inhibitory in PCR reactions, it is routinely added to restriction digests. The molecular weight is ∽66 kd. Acetylated BSA is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 mg/mL
Physical Form Liquid
Product Line Ambion
Quantity 6 x 25 mg
Packaging Tube

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.