Learn More
Invitrogen™ Cells-to-CT Bulk RT Reagents
Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription in the TaqMan™ Gene Expression Cells-to-CT™ Kit (20X RT enzyme mix and 20X RT buffer).
Brand: Invitrogen™ 4391852C
3880.19 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.12860kg
Includes: 20X RT Enzyme Mix, 2X RT Buffer
Description
Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription in the TaqMan™ Gene Expression Cells-to-CT™ Kit (20X RT enzyme mix and 20X RT buffer). These reagents are sufficient for 2500 RT reactions. When combined with the Cells-to- CT Bulk Lysis Reagent, these reagents can be easily incorporated into automated, high throughput applications.
Specifications
• 20X RT Enzyme Mix, Store at -20°C • 2X RT Buffer, Store at 4°C |
|
Fewer resources used and less waste, Less hazardous | |
cDNA Synthesis Reagents | |
20X |
Liquid | |
Cells-to-CT™ | |
Dry Ice | |
2500 reactions |
For Research Use Only. Not for use in diagnostic procedures.