Learn More
Invitrogen™ Cells-to-CT™ 1-Step TaqMan™ Kit
Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step TaqMan™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.
Brand: Invitrogen™ A25602
4229.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step TaqMan™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.
This new addition to the family includes a TaqMan™ 1-Step RT-PCR master mix. The single-tube format of the RT-PCR step streamlines the 'sample to answer' workflow more than ever. Combining the simple Cells-to- CT™ lysis technology with the fast cycling capability of the 1-step RT-PCR master mix allows quicker time to results with limited sample handling.
The Cells-to-CT™ 1-Step TaqMan™ Kit is:
- Complete—optimized workflow includes cell lysis reagents with gDNA removal and TaqMan™ 1-Step RT-PCR Master Mix
- Faster than ever—7-minute sample prep, including DNase treatment, combined with a 28-minute real-time RT-PCR step allows for ∼35 minute sample-to-answer workflow
- Simple—10 steps total (sample prep and real-time RT-PCR combined)
- Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
- Convenient—contains sufficient reagents to perform 5 real-time RT-PCRs for each cell sample you prepare; this allows you to study multiple targets or perform replicate RT-PCRs
Note: TaqMan™ primer/probe sets are sold separately.
Simple 7-minute Sample Preparation, Then Ready for Real-time RT-PCR
Whether you are using plates or tubes, the Cells-to-CT™ 1-Step TaqMan™ Kit uses a simple 7-minute sample preparation procedure. The lysis technology is designed for 10–100,000 cultured cells/sample. Cells are washed in PBS and lysed in solution for 5 minutes at room temperature with a simultaneous DNase treatment. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysate is now ready to analyze on your real-time PCR instrument. Simply add it to a real-time RT-PCR master mix containing the TaqMan™ primer/probe sets of interest. Alternatively, the lysate can be stored at –20°C. Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.
A Complete, Validated Solution
All components of the Cells-to-CT™ 1-Step TaqMan™ Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation and real-time 1-step RT-PCR master mixes. For added quality assurance, the Cells-to-CT™ 1-Step TaqMan™ Kit has been validated with TaqMan™ Gene Expression assays and shows performance equivalent to that obtained with purified RNA.
Accessory Product
The TaqMan™ Cells-to-CT™ Control Kit (Cat. No. 4386995) was designed for use with TaqMan™ Cells-to-CT™ products. This kit contains an endogenous control (ACTB) to normalize for sample loading variability and an artificial XenoRNA™ control and corresponding TaqMan™ Gene Expression assay to monitor the effects of PCR inhibition.
Specifications
Lysis Solution, 22 mL, store at 2°C to 8°C, DNase I, 220 μL, store at -5 to -30°C, Stop Solution, 2 x 1 mL, store at -5 to -30°C, TaqMan™ 1-step qRT-PCR Mix, 10 mL, store at -5 to -30°C, | |
Tube | |
1-step RT-qPCR | |
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, StepOne™ System, Real-Time PCR Systems or Thermal Cyclers, 7500 Fast Dx System, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system | |
Cells-to-CT™, TaqMan™ | |
400 preps | |
400 Reactions | |
Mammalian Cells | |
22 mL |
TaqMan | |
High | |
7 min. | |
Fewer resources used and less waste, Less hazardous | |
1-Step qPCR kit | |
Gene Expression | |
Cell Lysis | |
10-100,000 Cells |
Safety and Handling
- Cells-to-CT 1-Step TaqMan Kit
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Glycerin, Ethylene oxide (impurity), Glycerin, Dimethyl sulfoxide, Ethylene glycol, Tris (hydroxymet
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.