Learn More
Invitrogen™ Cells-to-Signal™ Lysis Buffer (10 ml Bottle)
The same lysis buffer found in the Invitrogen Cells-to-Signal™ Kit. Supplied in five 10 mL bottles.
Brand: Invitrogen™ AM8728
1045.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.09000kg
Description
The same lysis buffer found in the Invitrogen Cells-to-Signal™ Kit. Supplied in five 10 mL bottles.
Order Info
Shipping Condition: Room temperature
Specifications
Lysis Buffer may be stored at any temperature. | |
Cell Lysis Buffer | |
Room Temperature |
Ambion™, Cells-to-Signal™ | |
5 x 10 mL |
For Research Use Only. Not for use in diagnostic procedures.