missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellsDirect™ One-Step qRT-PCR Kit

The CellsDirect™ One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

Brand:  Invitrogen™ 11753100

552.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10043982

  • £650.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 10mL Resuspension Buffer; 1mL Lysis Enhancer; 500μL DNase I, Amplification Grade (1U/μL); 160μL 10X DNase I Buffer; 400μL 25mM EDTA; 100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL 50mM MgSO4; 2mL DEPC-treated water; 10μL HeLa Total RNA (10ng/μL); 100μL ROX Reference Dye
This item is not returnable. View return policy

Description

Description

The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

Efficiency: offers significant time and cost savings for increased throughput
Convenience: enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
Sensitivity: no separate RNA isolation step increases sensitivity by minimizing sample loss
Broad detection range: detect targets from samples ranging from a single cell to 10,000 cells
Versatility: compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)

Streamline your workflow
In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.

Ideal for your qRT-PCR application
CellsDirect technology is validated for gene expression studies involving limited samples. CellsDirect One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps. The CellsDirect One-Step qRT-PCR Kit may also be used to detect genomic DNA targets. CellsDirect One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g., TaqMan™ or LUX™ fluorogenic primers).

This kit includes a separate tube of ROX™ Reference Dye. An equivalent kit with ROX Reference Dye included in the 2X Reaction Mix is also available.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10 mL Resuspension Buffer
• 1 mL Lysis Enhancer
• 500 μL DNase I, Amplification Grade (1 U/μL)
• 160 μL 10X DNase I Buffer
• 400 μL 25 mM EDTA
• 100 μL SuperScript™ III RT/Platinum™ Taq Mix
• 2 × 1.25 mL 2X Reaction Mix
• 1 mL 50 mM MgSO4
• 2 mL DEPC-treated water
• 10 μL HeLa Total RNA (10 ng/μL)
• 100 μL ROX™ Reference Dye (store at -20°C in the dark)
The kit is shipped on dry ice and should be stored at -20°C
Fast or Standard
CellsDirect™, Platinum™, SuperScript™
2X
Cells
Primer-probe
7500 Fast System, 7500 System, BioRad iQ5, BioRad MyiQ, Corbett RotorGene, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad iCycler iQ, Cepheid SmartCycler, MJ Chromo4, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system
RT-PCR
100 reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.