Learn More
Invitrogen™ Conical Tubes (50 mL) (bagged)
These polypropylene tubes are sterile, nuclease-free, nonpyrogenic, and can be autoclaved to 121°C.
Brand: Invitrogen™ AM12502
Code : 50
Additional Details : CAS Number : 9048-46-8 Weight : 0.36000kg
Description
- Provided assembled with green screw caps
- The tubes have a printed writing space and volume marks on the sidewalls for convenience
- These polypropylene tubes are sterile, nuclease-free, nonpyrogenic, and can be autoclaved to 121°C
- 200 x 50 mL tubes
Order Info
Shipping Condition: Room Temperature
Specifications
50 mL | |
Sterile | |
Ambion™ | |
Yes | |
Autoclavable | |
Conical | |
Yes | |
Conical Tube |
Polypropylene | |
Store at room temperature. | |
Room Temperature | |
Yes | |
Conical | |
Screw Cap | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.