Learn More
Invitrogen™ DEPC-Treated Water
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC.
Brand: Invitrogen™ 4387937
114.00 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Description
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Specifications
6 to 8 | |
Bottle | |
4 x 1000 mL | |
Ambion™ |
Molecular Biology | |
DEPC-Treated | |
Autoclaved, Membrane-Filtered |
For Research Use Only. Not for use in diagnostic procedures.