Learn More
Invitrogen™ DNAclear™ Purification Kit
Designed to purify double-stranded DNA (dsDNA) generated by enzymatic reactions such as cDNA synthesis and PCR
Brand: Invitrogen™ AM1756
Code : 50
Additional Details : Weight : 0.22000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Description
The DNAclear™ cDNA Purification Kit process is simple and fast. DNA amounts ranging from 1ng to 3μg can be efficiently purified and concentrated. The DNAclear procedure consists of three steps: 1) DNA is bound to the Microfilter Cartridge in the presence of the DNA Binding Buffer, 2) the DNA is washed with the cDNA Wash Buffer, and 3) the DNA is eluted with preheated nuclease-free water. The DNAclear Kit can be used to remove free nucleotides, oligonucleotides, proteins, and salts from dsDNA samples. The DNA that is recovered from the Microfilter Cartridge is ready for any application that requires highly pure DNA including in vitro transcription reactions.
- Includes sufficient reagents for 20 purifications
- Efficiently purify and concentrate dsDNA ranging from 1ng to 3μg
- Ideal for downstream applications such as in vitro transcription
- Simple and fast protocol: about 10 minutes
DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up
Order Info
Shipping Condition: Room Temperature
Specifications
cDNA Purification Kit | |
cDNA Binding Buffer, cDNA Wash Buffer, Microfilter Cartridges + Collection Tubes, and Elution Tubes should be stored at room temperature. Nuclease-free water may be stored at any temperature. | |
Not High-throughput Compatible (Manual) | |
Room Temperature | |
Ambion™, DNAclear™ | |
20 Preps |
Centrifuge | |
DNA | |
PCR Products | |
Microtube | |
PCR, Sequencing, In Vitro Transcription, Cloning |
For Research Use Only. Not for use in diagnostic procedures.