missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ mRNA DIRECT™ Micro Purification Kit

Product Code. 10531075
Click to view available options
Quantity:
2 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10531075

Brand: Invitrogen™ 61021

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2mL Beads and Buffers

The Dynabeads mRNA DIRECT Micro Kit isolates highly purified and intact mRNA directly from cells, tissue, and total RNA samples.

The Dynabeads mRNA DIRECT Micro Kit isolates highly purified and intact mRNA directly from cells, tissue, and total RNA samples.

  • Obtain extremely pure mRNA in about 15 minutes
  • Integrate total RNA purification and mRNA enrichment steps
  • Perform reverse transcription and PCR amplification right on the bead
  • Use in whole transcriptome library preparation with the Ion Total RNA-Seq Kit v2.0
  • Final Product: mRNA

Not high-throughput compatible (Manual)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 mL Beads and Buffers

Store at 2°C to 8°C.
Isolation Technology Magnetic Bead
Sample Type Liquid Samples (e.g. Serum), Plant Samples, Viral Samples, Cells, Tissue, FFPE and Fixed Samples, Yeast, RNA, Blood
Final Product Type mRNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Reverse Transcriptase PCR (RT-PCR)
Product Line DYNAL, Dynabeads, mRNA DIRECT
Product Type Micro Purification Kit
Quantity 2 mL
Shipping Condition Room Temperature
Target mRNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.