missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDTA (0.5 M), pH 8.0, RNase-free

Product Code. 10135423
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
100mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10135423

Brand: Invitrogen™ AM9260G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.

Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
Content And Storage Store at room temperature.
Concentration 0.5 M
Packaging Type Bottle
pH 8
Product Line Ambion
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid
compliance-icons
Product Identifier
  • 0.5M EDTA PH 8.0 100ML
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Glycine, N,N-1,2-ethanediylbis[N-(carboxymeth yl)-, disodiumsalt, dihydrate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.