missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Exo-Klenow Fragment (cloned), DNA Polymerase I, 5U/μL

No exonuclease activity

Brand:  Invitrogen AM2008

Product Code. 10196904

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: One tube containing 750U (5U/μL)
This item is not returnable. View return policy

Description

Description

  • Ambion Exonuclease-free Klenow Fragment is a genetically engineered Klenow polymerase to remove 3' to 5' exonuclease activity has been removed, leaving only the 5' to 3' polymerase activity
  • One tube containing 750U (5U/μL) is provided
  • The elimination of exonuclease activity makes Exo-Klenow Fragment the enzyme of choice for isotopic or biotin labeling of DNA probes by the random-primed method, and for DNA sequencing by the Sanger dideoxy method. The Exo-Klenow Fragment is thoroughly tested for nonspecific contaminating endonuclease, exonuclease, and protease activity
  • Functionality is determined with a random-priming reaction
  • Unit Definition: One unit of enzyme incorporates 10nmol of deoxynucleotides into acid insoluble material in 30 minutes at 37°C

Cloning, Restriction Enzyme Cloning

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

One tube containing 750U (5U/μL). Store at -20deg.C
750 units
DNA Sequencing
5U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.