missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)

Product Code. p-4926963
Click to view available options
Quantity:
300 μL
5 x 300 μL
Unit Size:
1500µL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10301575

Brand: Invitrogen™ AM9516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant.

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.

Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 15 mg/mL
Content And Storage Store at –20°C.
Packaging Type Tube
Product Line Ambion, GlycoBlue
Purity Molecular Biology Grade
Quantity 5 x 300 μL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.