Learn More
Invitrogen™ E. coli Total RNA
The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200 μg is provided at a concentration of 1 mg/mL.
Marke: Invitrogen™ AM7940
301.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Weitere Details : Weight : 0.10500kg
Beschreibung
The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200 μg is provided at a concentration of 1 mg/mL. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Because it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.
Bestellinformation
Shipping Condition: Dry ice
Spezifikation
RNA: varies | |
200 μg E. coli Total RNA; store at or below –70°C |
|
E. coli total RNA stored in THE RNA Storage Solution (1 mM Sodium Citrate, pH 6.4; Cat No. AM7000) | |
200 μg | |
Dry Ice |
1 mg/mL | |
E. coli total RNA isolated from DH5α cells | |
RT-PCR, ribonuclease protection assays, S1 nuclease assays, cDNA synthesis, Northern analysis, IVT | |
DNase-Treated |
For Research Use Only. Not for use in diagnostic procedures.