missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ In Vivo Pre-Designed siRNA

Product Code. 15286304
Click to view available options
Final Product Type:
Ambion™ In Vivo Custom SIRNA HPLC
Ambion™ In Vivo Pre-Designed HPLC IVR
Ambion™ In Vivo Pre-Designed SIRNA HPLC
Ambion™ In Vivo Pre-Designed siRNA
Quantity:
1 μmol
250 nmol
Unit Size:
250µmol
Each
This item is not returnable. View return policy

Product Code. 15286304

Brand: Invitrogen™ 4457308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models.

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models. These siRNAs are chemically modified to enhance stability, reduce immunogenicity, and improve delivery efficiency, making them excellent for in vivo studies. Ambion In Vivo Predesigned siRNAs allow researchers to investigate gene function in a physiological context, offering valuable insights into gene regulation mechanisms and disease processes. Built using the Silencer Select algorithm, these predesigned siRNAs ensure high specificity and efficiency.

Key features include:

  • Enhanced stability—chemical modifications help ensure that Ambion In Vivo Predesigned siRNAs remain stable in the bloodstream and within tissues, helping provide sustained gene silencing effects
  • Reduced immunogenicity—modifications minimize immune responses, helping ensure safe and effective use in animal models
  • Efficient delivery—optimized for use with various delivery systems, including lipid nanoparticles, helping ensure efficient uptake by target cells
  • High specificity—designed to precisely target specific mRNAs, resulting in potent and specific gene knockdown

In vivo-ready siRNA

Ambion In Vivo Predesigned siRNAs are processed to be ready for in vivo use. This includes HPLC purification, membrane dialysis to remove salts and solvents, and sterile filtration prior to drying down.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Description Ambion™ In Vivo Pre-Designed siRNA, 250nmol, HPLC, IVR, Predesigned, Incorporate the Latest Improvements In siRNA Design, Guaranteed-to-silence Based on Their Proven Design
RNAi Type siRNA
Final Product Type Ambion™ In Vivo Pre-Designed siRNA
Product Line Ambion
Product Type Ambion™ In Vivo Pre-Designed siRNA
Purity Standard
Quantity 250 nmol
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.