missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Ambion™ In Vivo Pre-Designed siRNA

Product Code. 15591104
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15591104

Brand: Ambion™ 4457298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models.

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models. These siRNAs are chemically modified to enhance stability, reduce immunogenicity, and improve delivery efficiency, making them excellent for in vivo studies. Ambion In Vivo Predesigned siRNAs allow researchers to investigate gene function in a physiological context, helping provide valuable insights into gene regulation mechanisms and disease processes. Built using the Silencer Select algorithm, these predesigned siRNAs help ensure high specificity and efficiency.

Ambion™ siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. These siRNAs are guaranteed-to-silence based on their proven design.

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store in freezer (-5°C to -30°C).
Description Ambion™ In-vivo Pre-designed siRNA, 5nmol, Powder, White, Standard Purity, Combined Knockdown Effectiveness and Stability, Easy Tracking of Administered siRNAs, Double-Stranded siRNAs That are Recognized by the RNA-induced Silencing Complex (RISC) to
RNAi Type siRNA
Final Product Type Ambion™ In Vivo Pre-Designed siRNA
Product Line Ambion
Product Type Ambion™ In-vivo Pre-designed siRNA
Purity Standard
Quantity 5 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.