missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Power SYBR™ Green Cells-to-CT™ Kit
GREENER_CHOICE

Product Code. 10259494
Click to view available options
No. of Reactions:
100 reactions
40 reactions
400 reactions
Unit Size:
100 reactions
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10259494

Brand: Invitrogen™ 4402955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ambion™ Power SYBR™ Green Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results.

Ambion Power SYBRGreen Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results. A breakthrough cell lysis and RNA stabilization technology eliminates the laborious and time-consuming RNA purification process completely. Power SYBRGreen Cells-to-CT Kit is Powered by AmpliTaq Gold™ DNA Polymerase, LD, which provides the highest levels of specificity with standard real-time PCR.

  • Includes optimized reagents to work efficiently, right out of the box
  • Simple, effective Cells-to-CT technology enables sample preparation at room temperature in only 7 minutes, including DNase treatment
  • Validated accuracy, exceptional reproducibility, and maximal sensitivity from 10 to 100,000 cells per sample; results equivalent to those from purified RNA

Extraordinary Value
The kit includes reagents for sample preparation integrated into a complete, optimized gene expression workflow including reverse transcription reagents and a high-performance PCR master mix with SYBRGreen.

Extraordinary Ease
The protocol begins with a simple 7-minute sample preparation procedure. First, cells are washed in PBS, and then lysed for 5 minutes at room temperature; DNase treatment can be performed concurrently. The lysis reaction is stopped, and the sample is then ready for reverse transcription or storage at -20°C for up to 5 months. Because samples are processed directly in culture plates (96- or 384-well), sample handling and the potential for sample loss or transfer error are minimized, resulting in higher reproducibility. Following sample preparation, a portion of the cell lysate is added to an RT reaction followed by real-time PCR using the included master mix.

Extraordinary Performance
To demonstrate the performance of the kit, replicate experiments were run using either traditional RNA purification and real-time PCR analysis, or the Power SYBRGreen Cells-to-CT Kit. The kit shows equivalent performance to results obtained using purified RNA for analysis of 10 to 100,000 cells. In addition, the sensitivity, efficiency, and dynamic range were shown to be superior to competitor lysate kits. Samples prepared with the kit have been tested for compatibility on various real-time PCR platforms including those from Applied Biosystems, Roche, and Bio-Rad.

  • PCR Method: 2-Step qRT-PCR
  • Purity/Quality Grade: LD (Low DAN)
  • Passive Reference Dye: ROX (Pre-mixed)
  • Polymerase: AmpliTaq Gold™ DNA Polymerase

Accessory Product:
The SYBRGreen Cells-to-CT Control Kit (Cat. No. 4402959) contains a Xeno™ RNA control and primer set and is recommended to monitor the efficiency of cell lysis and amplification inhibition in Cells-to-CT reactions.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Content And Storage Store Stop Solution, DNase I and 20X RT Enzyme Mix at -20°C. Store Lysis Solution, 2X SYBR™ RT Buffer, PowerSYBR™ Green PCR Master Mix at 4°C. Store Nuclease-free Water at room temperature
Detection Method SYBR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7200 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 5700, GeneAmp 9600, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Green Features Fewer resources used and less waste, Less hazardous
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line Cells-to-CT, SYBR
Purification Time 5 min.
Purity or Quality Grade LD (Low DNA)
Quantity 400 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Hot Start Built-In Hot Start
No. of Reactions 400 reactions
Sample Type Cells
Show More Show Less
compliance-icons
Product Identifier
  • ower SYBR Green Cells-to-CT Kit
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase, tritirachium album serine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.