missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Ambion™ In Vivo Pre-Designed siRNA
Ambion™ In Vivo Pre-Designed siRNA
£1842.00 - £3670.00
Specifications
Content And Storage | Room temperature |
---|---|
Product Type | Custom Ambion™ In-vivo Pre-designed siRNA |
Purity | HPLC In-vivo Ready |
Form | Solid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15531114
|
Ambion™
4457307 |
250 nmol |
£1842.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15541114
|
Ambion™
4457309 |
1 umol |
£3670.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.Specifications
Room temperature | |
Custom Ambion™ In-vivo Pre-designed siRNA | |
HPLC In-vivo Ready | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.