Learn More
Invitrogen™ IPTG (Dioxane-Free)
Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage.
£113.00 - £213.00
Specifications
Chemical Name or Material | IPTG (Isopropyl β-D-thiogalactopyranoside) |
---|---|
Content And Storage | Store at –20°C. |
Packaging Type | Bottle |
Product Line | Ambion™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10685985
|
Invitrogen™
AM9464 |
5 g |
£213.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10556735
|
Invitrogen™
AM9462 |
2 g |
£113.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon.
Order Info
Shipping Condition: Room temperature
Specifications
IPTG (Isopropyl β-D-thiogalactopyranoside) | |
Bottle | |
Room Temperature |
Store at –20°C. | |
Ambion™ | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.