missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LeukoLOCK™ Fractionation & Stabilization Kit with Manual

Product Code. 15544394
Click to view available options
Product Type:
Kit
Kit with manual
Unit Size:
Each
Pack of 20
This item is not returnable. View return policy

Product Code. 15544394

Brand: Invitrogen™ AM1933M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

RNAlater Solution, Rubber Septum Caps, White Slip Connectors, Transfer Spikes, LeukoLOCK Filters, and Elution Solution, and 1X PBS

The Ambion™ LeukoLOCK™ Fractionation & Stabilization Kit is one of two kits that comprise the Total RNA Isolation System (patent pending), an innovative method for cellular fractionation of whole blood and total RNA stabilization and extraction from the leukocyte population.

The Ambion™ LeukoLOCK™ Fractionation & Stabilization Kit is one of two kits that comprise the Total RNA Isolation System (patent pending), an innovative method for cellular fractionation of whole blood and total RNA stabilization and extraction from the leukocyte population. The kit includes sufficient reagents for 20 reactions. The LeukoLOCK™ System (Cat. No. AM1923) is optimized for use with human blood. Blood is a storehouse of cellular information; however, the presence of globin mRNA in RNA prepared from whole blood can interfere with downstream expression profiling applications. The LeukoLOCK™ System employs filter-based leukocyte-depletion technology to isolate leukocytes from whole blood and Ambion's RNAlater™ Solution to stabilize the cells on the filter. By excluding red blood cells, the RNA that is purified from captured leukocytes is inherently depleted of globin mRNA, which improves sample utility for expression profiling and other applications (see Figure).

LeukoLOCK™ Fractionation & Stabilization Kit
A 9–10 mL sample of anticoagulated blood is passed through a LeukoLOCK™ filter, which captures the total leukocyte population while eliminating red blood cells (including reticulocytes), platelets, and plasma. After rinsing with phosphate-buffered saline, the filter is flushed with RNAlaterSolutionto stabilize the RNA in the captured leukocytes (see protocol schematic). The RNA can be isolated immediately, or stabilized cells can be maintained for several days at room temperature or for longer periods at –20°C or –80°C (see Figure).

Accessory Products:
LeukoLOCK™ Fractionation & Stabilization Kit is designed for use with the LeukoLOCK™ Total RNA Isolation Kit (Cat. No. AM1934). These kits are offered separately to accommodate blood sample collection at one location and sample processing at a different location, but may be purchased together as the LeukoLOCK™ Total RNA Isolation System (Cat. No. AM1923).

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Content And Storage The RNAlater™ Solution, Rubber Septum Caps, White Slip Connectors, Transfer Spikes, LeukoLOCK™ Filters, and Elution Solution, should be stored at room temperature. 1X PBS may be stored at any temperature.
Isolation Technology Vacuum Filter Column, Magnetic Bead
Sample Type Blood
Final Product Type Cells (Leukocytes)
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Northern Blotting
Includes RNAlater solution, rubber septum caps, white slip connectors, transfer spikes, LeukoLOCK filters, elution solution, 1X PBS, manual
Prep Scale > 1 mL Blood Samples (Large-scale)
Product Line Ambion, LeukoLOCK
Product Type Kit with manual
Target Total RNA
Yield 20 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.