Learn More
Invitrogen™ LiCl Precipitation Solution (7.5 M)
Precipitation solution offers major advantages over other RNA precipitation methods in that it does not precipitate DNA, protein, or carbohydrates.
Brand: Invitrogen™ AM9480
89.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : CAS Number : 7447-41-8 Weight : 0.10500kg
Description
Precipitation solution offers major advantages over other RNA precipitation methods in that it does not precipitate DNA, protein, or carbohydrates. It is supplied in one bottle of 100 mL at a concentration of 7.5M. It is the method of choice for removing inhibitors of translation or cDNA synthesis from RNA preparations.
Note: LiCl precipitation may not remove all DNA contamination from total RNA isolations. If the RNA is to be used for RT-PCR, it should be DNase I treated. The solution is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
7.5 M | |
RNA Extraction | |
DNase-Free, RNase-Free | |
Room Temperature |
Store at 2°C to 8°C. | |
Ambion™ | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.