missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagMAX™-96 Blood RNA Isolation Kit

Product Code. 10743077
Click to view available options
Quantity:
96 preps
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10743077

Brand: Invitrogen™ AM1837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

This kit is for the rapid, high-throughput purification of pathogen RNA from mammalian whole blood, milk, and other biofluids in 96-well plates.

This Ambion™ kit is for the rapid, high-throughput purification of pathogen RNA from mammalian whole blood, milk, and other biofluids in 96-well plates. RNA purified using the kit is ideal for RT-PCR applications. The kit uses magnetic bead–based purification and includes sufficient reagents for 96 reactions.

• Easily isolate viral or total RNA from whole blood
• Save precious time—complete the protocol in less than 1 hour
• Walk away—integrate with established robotic platforms
• Validated with human, bovine, porcine, and ovine whole blood

Benefits of MagMAX™ Magnetic Bead-Based Purification:
Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high- efficiency binding, the bound RNA can be eluted in just 30–50 μL of nuclease-free water, concentrating RNA from large, dilute samples.

Isolate RNA from as Little as 50 μl of Blood:
For the highest-quality RNA, use whole blood samples that have not been frozen. Approximately 300–500 ng of intact RNA can be consistently purified from 50 μl of human whole blood. Bovine, porcine, and ovine whole blood samples can also be used. Yields vary from species to species (see data). The recovered RNA is devoid of PCR inhibitors such as proteins and heme, commonly found in RNA isolated from blood.

Isolate Viral and Total RNA from Whole Blood Samples:
The MagMAX™-96 Blood RNA Isolation Kit eliminates sample preprocessing, which can cause sample mix-up and cross-contamination. The procedure is fast (less than 1 hour), simple, and well suited for automation. The kit can also be used to process fewer than 96 samples at one time and is compatible with either manual processing using multi-channel pipettors or processing with robotic liquid handlers. Downloadable protocols for the use of this kit with specific liquid handling systems are available at our Automation Resource. The MagMAX™-96 Blood RNA Isolation Kit can also be used for total nucleic acid isolation by omitting the DNase treatment.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Processing Plate with Lid, Lysis/Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, and Elution Buffer at room temp.

Store MagMax Turbo DNase Buffer and RNA Binding Beads at 4°C.

Store Lysis/Binding Enhancer and TURBO DNase at -20°C.
Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Liquid Samples (e.g. Serum), Milk, Viral Samples, Whole Blood (Mammalian)
Final Product Type Total RNA, Viral RNA
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
For Use With (Equipment) MagMAX™ Express 24 and 96, Robotic Liquid Handling Systems, MagMAX™ Express-96 Standard Magnetic Particle Processor
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions 96 Preps
Prep Scale < 100 μL Blood Samples (Small-scale)
Product Line Ambion, MagMAX
Quantity 96 preps
Starting Material Amount Up to 50 μl whole blood
Target Viral DNA/RNA, Total RNA
Yield 0.5 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.