Learn More
Invitrogen™ MagMAX™ Wash Solution 2 Concentrate
Invitrogen MagMAX Wash Solution 2 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately.
Brand: Invitrogen™ AM8640
Code : 50
Additional Details : Weight : 0.02000kg
Description
Invitrogen MagMAX Wash Solution 2 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately. Supplied in one 200 mL bottle.
Order Info
Shipping Condition: Room temperature
Specifications
Wash Buffer | |
Store MagMAX™ Wash Solution 2 Concentrate at 4°C or room temperature. | |
Ambion™, MagMAX™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.