missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAclear™-96 Transcription Clean-Up Kit

Quickly cleanup large-scale transcription reactions

Brand:  Invitrogen™ AM1909

Product Code. 10127304

  • £376.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 8mL Binding Solution, 20mL Wash Solution Concentrate, 10mL Elution Solution, 20mL Nuclease-free Water, 1 MEGAclear-96 Filter Plate, 1 Collection Plate, 2 Adhesive Plate Sealers
This item is not returnable. View return policy

Description

Description

MEGAclear-96 Kit is designed for quick, high-throughput cleanup of any large-scale transcription reaction. The kit efficiently separates RNA from unincorporated NTPs, enzymes, and buffer components.

  • Purifies transcription reactions in a 96-well format to produce high quality RNA free of unincorporated NTPs, enzymes, and buffer components
  • Quick protocol—purify RNA in less than 20 minutes
  • Can be adapted to robotic platforms
  • Protocol is quick and easy
  • Can also be integrated with your robotic platform or used with manual methods

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 8 ml Binding Solution (store at 4°C)
• 20 ml Wash Solution Concentrate (store at 4°C)
• 10 ml Elution Solution (store at room temperature, 4°C, or -20°C)
• 20 ml Nuclease-free Water (store at room temperature, 4°C, or -20°C)
• 1 MEGAclear™-96 Filter Plate (store at room temperature)
• 1 Collection Plate (store at room temperature)
• 2 Adhesive Plate Sealers (store at room temperature)
96-well Plate
mRNA, Synthetic mRNA
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
High-throughput Compatible
Ambion™, MEGAclear™
96 Preps
Room Temperature
100 μg
Purified Synthetic mRNA, Purified mRNA
Silica Spin Column
mRNA, Synthetic mRNA
Vacuum Manifold, Centrifuge
96 Preps
mRNA purification kit, Purification kit
30 min
1 ng to 500 μg of dsRNA>200b or ssRA>100bp
synthetic mRNA, mRNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.