missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAscript™ T7 Transcription Kit

Ultra-high yield technology for in vitro transcription

Brand:  Invitrogen™ AM1333

234.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 50

Additional Details : Weight : 0.03000kg


 View more versions of this product

Product Code. 10629645

  • £252.00 / 25 reactions
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

The high yields are achieved by modifying typical transcription reaction conditions so that very high nucleotide concentrations can be effectively used. The T7 Enzyme Mix and the 10X Reaction Buffer are specifically calibrated for each lot and RNA polymerase.

Using the MEGAscript T7 Kit:
The MEGAscript T7 Kit contains in vitro transcription reaction components and a control template. The kit will yield a total of 3 to 5mg of RNA (approximately 100μg of RNA or more per reaction) from the control template supplied with the kit. This corresponds to 400 to 650 moles of RNA for each mole of template. Smaller templates typically yield a lower mass and a higher molar yield of product. Novel transcription reaction conditions and a patented, high-yield technology allow the synthesis of 10 to 50 times the amount of RNA produced by conventional transcription reactions.

Applications:The MEGAscript T7 Kit is intended for the synthesis of large amounts of unlabeled or low specific activity RNA for a variety of uses, including in vitro translation, antisense/microinjection studies, and isolation of RNA binding proteins. In large-scale transcription reactions, the concentration of all four nucleotides is high, well above the Km for the enzyme. The MEGAscript T7 Kit typically yields over 10 times more RNA than conventional in vitro transcription reactions. The kit is not recommended for synthesis of high specific activity probes.

Accessory products:
MessageAmp™ aRNA amplification kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use MEGAclear™ Kit (Mfr. No. AM1908) and the MEGAclear-96 Kit (Mfr.. No. AM1909).

RNA Yield:
100 μg

Specifications

Specifications

T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Kit
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
Ambion™, MEGAscript™
25 Reactions
Dry Ice
PCR Products, Linearized DNA plasmid
Synthetic mRNA, mRNA
mRNA, Synthetic mRNA
25 Reactions
T7 In Vitro Transcription kit
T7
100 μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MEGASCRIPT T7 KIT 25 RXNS EACH

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formamide, lithium chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.