missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MessageAmp™ II-Bacteria RNA Amplification Kit

For highly sensitive, whole-genome expression analysis

Brand:  Invitrogen™ AM1790

Product Code. 10259724

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The kit uses an in vitro transcription (IVT)–mediated linear amplification method optimized for use with bacterial RNA.

Using the MessageAmp II-Bacteria RNA Amplification Kit
This kit is the only commercially available kit for amplification of prokaryotic mRNA. The kit is compatible with any bacterial species and input RNA prepared from multiple sources, such as total RNA, bacterial mRNA or RNA. aRNA generated by the MessageAmp II-Bacteria Kit can be labeled by either inclusion of a labeled nucleotide in the IVT reaction, or by reverse transcription of the aRNA in a reaction containing a labeled nucleotide. The MessageAmp II-Bacteria Kit can thus be used to prepare both 33P-labeled sense strand cDNA (for nylon PCR arrays) and biotin- or fluorescent dye–labeled aRNA for glass slide microarrays or Affymetrix GeneChip™ arrays.

Accessory products
Bacterial mRNA can be enriched from total RNA using the MICROBExpress™ Bacterial mRNA Enrichment Kit (Cat. No. AM1905). Bacterial RNA can be separated from host cell mixtures with the MICROBEnrich™ Kit (Cat. No. AM1901).

  • Maximum sensitivity: amplify as little as 10ng of mRNA or 100ng total RNA; a 10- to 500-fold reduction in the input RNA needed to probe microarrays
  • Highest yield of full-length transcripts: includes ArrayScript™ RT which produces up to twice the amount of full-length cDNA as M-MLV, AMV, or SuperScript™ II
  • Stringently reliable data: reactions are highly reproducible, with Pearson's Correlation Coefficients >0.995

Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bacteria RNA Amplification Kit
Store the following at -20°C in a frost-free freezer:
• 22 μl PAP
• 22 μl 10X Poly(A) Tailing Buffer
• 12 μl Poly(A) Tailing ATP
• 22 μl T7 Oligo(dT) VN Primer
• 22 μl ArrayScript™ Reverse Transcriptase
• 22 μl RNase Inhibitor
• 22 μl 10X First Strand Buffer
• 170 μl dNTP Mix
• 210 μl 10X Second Strand Buffer
• 42 μl DNA Polymerase
• 22 μl RNase H
• 84 μl T7 Enzyme Mix
• 84 μl 10X T7 Reaction Buffer
• 84 μl each T7 ATP, CTP, GTP and UTP
• 1.75 ml Nuclease-free Water
• 10 μl Control RNA (1 mg/ml)

Store the following at room temperature:
• 30 ml Wash Buffer
• 7 ml cDNA Binding Buffer
• 9 ml aRNA Binding Buffer
• 20 aRNA Filter Cartridges
• 40 aRNA Collection Tubes
• 20 cDNA Filter Cartridges + Tubes
• 20 cDNA Elution Tubes• 10 ml Nuclease-free Water
ArrayScript™
Kit
20 Reactions
Prokaryotic Cell
No
mRNA (Bacterial), Total RNA (Bacterial)
Ambion™, MessageAmp™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.