Learn More
Invitrogen™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit
Generate RNA transcripts with higher protein yields
Brand: Invitrogen™ AM1345
452.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.06000kg
Description
The mMESSAGE mMACHINE T7 Ultra Kit combines new cap analog, anti-reverse cap analog (ARCA), with patented high-yield transcription technology to generate RNA transcripts that produce higher protein yields compared to other transcripts upon translation.
- RNA Yield: 20-30μg
- Synthesize more protein from in vitro-transcribed RNA
- Express RNA transcripts more efficiently both in vitro and in vivo
- Stabilize transcripts in vivo with included reagents for poly(A) tailing
Specifications
T7 Enzyme Mix, 10X Reaction Buffer 2XNTP⁄ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Formaldehyde Load Dye are all stored at –20°C. Nuclease-free Water may be stored at any temperature. | |
Kit | |
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products | |
Ambion™, mMESSAGE mMACHINE™ | |
10 Reactions | |
Dry Ice | |
PCR Products, Linearized DNA plasmid |
Synthetic mRNA, mRNA, ARCA Capped mRNA | |
mRNA, Synthetic mRNA | |
10 Reactions | |
T7 In Vitro Transcription kit | |
T7 | |
20-30 μg |
For Research Use Only. Not for use in diagnostic procedures.