Learn More
Invitrogen™ Max Bacterial Enhancement Reagent
The Max Bacterial Enhancement Reagent is designed for use with TRIzol™ Reagent to improve the isolation of intact total RNA up to three-fold from Gram-positive and Gram-negative bacteria.
Brand: Invitrogen™ 16122012
316.68 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Includes: Max Bacterial Enhancement Reagent
Description
The Max Bacterial Enhancement Reagent is designed for use with TRIzol™ Reagent to improve the isolation of intact total RNA up to three-fold from Gram-positive and Gram-negative bacteria. The Max Bacterial Enhancement Reagent is a ready-to-use solution composed of chelating agents, detergent, and a buffer, and is used as an efficient pre-treatment buffer for bacterial cells prior to RNA isolation with TRIzol Reagent. The use of Max Bacterial Enhancement Reagent with TRIzol inactivates endogenous RNases and promotes protein denaturation, improving the RNA quality and integrity. The Max Bacterial Enhancement Reagent is available either separately or packaged with TRIzol as part of the TRIzol Max™ Bacterial RNA Isolation Kit (Cat. No. 16096020 or 16096040). 100 preps. Manual; not high-throughput compatible.
Specifications
Includes Max Bacterial Enhancement Reagent. Store at room temperature. | |
Not High-throughput Compatible (Manual) | |
1 Ea. | |
miRNA analysis | |
Organic Extraction | |
Total RNA, Transcriptome RNA, miRNA | |
20 mL |
TRIzol Reagent | |
DNase-Free, RNase-Free | |
Scaleable | |
Total RNA, Transcriptome RNA, micro RNA | |
100 Reactions | |
Bacteria (Gram-Negative), Bacteria (Gram-Positive) |
For Research Use Only. Not for use in diagnostic procedures.