missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse RiboPure™-Blood RNA Isolation Kit with Manual

For the purification of extremely high-quality total RNA (and optionally, microRNA) from mouse or rat blood in about 35 minutes.

Brand:  Invitrogen™ AM1951M

 View more versions of this product

Product Code. 10584825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Acid–Phenol:Chloroform, 3M Sodium Acetate pH 5.5, Wash Solution 1 Concentrate, Wash Solution 2/3 Concentrate, 2mL tubes pre-loaded with RNAlater, Lysis Solution, Filter Cartridges, Collection Tubes, Nuclease–free Water and Elution solution
This item is not returnable. View return policy

Description

Description

For the purification of extremely high-quality total RNA (and optionally, microRNA) from mouse or rat blood in about 35 minutes. The Ambion™ kit includes sufficient reagents for 25 RNA purifications from whole blood samples ranging from 0.1–0.5 each.

• Includes RNAlater™, predispensed in 2 mL tubes, for RNA stabilization
• Resulting RNA is suitable for stringent downstream applications such as array analysis and qRT-PCR

Due to interference from the enzymatic inhibitors and anticoagulants present in blood samples, RNA purification from whole blood requires a stringent isolation method that yields high-quality RNA depleted of inhibiting agents and contaminants. The kit includes microfuge tubes preloaded with RNAlater™ Tissue Collection:RNA Stabilization Solution, an aqueous, nontoxic storage reagent that prevents RNA degradation and preserves RNA profiles within samples. Stabilized samples can be stored at room temperature for up to 3 days or at –20°C for long-term storage. The related Figures show the performance of RNA isolated with the Mouse RiboPure™-Blood Kit in real-time RT-PCR. The kit includes an alternative protocol for isolating total RNA that contains microRNA. Typically, 30–55 μg of total RNA (containing microRNA) is obtained from 0.5 mL mouse blood. Yields from rat blood are considerably more variable, generally 18–40 μg from 0.5 mL blood.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acid-Phenol:Chloroform, 3M Sodium Acetate pH 5.5, Wash Solution 1 Concentrate, Wash Solution 2/3 Concentrate at 4°C. Store 2 mL tubes pre-loaded with RNAlater, Lysis Solution, Filter Cartridges, Collection Tubes, Nuclease-free Water and Elution solution at room temperature.
Organic Extraction, Spin Column
Mouse
Total RNA, micro RNA
Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
Ambion™, RiboPure™
Up to 0.5 ml whole blood
4 μg
Filter Cartridge, Tube
35 min.
Blood
Not High-throughput Compatible (Manual)
Acid–Phenol:Chloroform, 3M sodium acetate pH 5.5, wash solution 1 concentrate, wash solution 2/3 concentrate, 2mL tubes pre-loaded with RNAlater, lysis solution, filter cartridges, collection tubes, nuclease-free water, manual
Kit with manual
miRNA, Total RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.