Learn More
Invitrogen™ NaCl (5 M), RNase-free
Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.
Brand: Invitrogen™ AM9760G
Description
Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature

Specifications
Sodium Chloride | |
5 M | |
Ambion™ | |
100 mL | |
Liquid |
Store at room temperature. | |
Bottle | |
Molecular Biology Grade | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2858739 | Certificate of Analysis | 28/01/2025 | AM9760G |
2652656 | Certificate of Analysis | 28/01/2025 | AM9760G |
2865040 | Certificate of Analysis | 28/01/2025 | AM9760G |
2877507 | Certificate of Analysis | 28/01/2025 | AM9760G |
2890191 | Certificate of Analysis | 28/01/2025 | AM9760G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.