Learn More
Invitrogen™ NorthernMax™ 10X Running Buffer
For use in denaturing formaldehyde agarose gel systems
Brand: Invitrogen™ AM8671
Code : 50
Additional Details : Weight : 0.10500kg
Description
- Ready to use and certified Rnase-free
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- Compatible with agarose gels
- Comprehensive instructions are included
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 4°C. | |
10X | |
Agarose Gels |
Running Buffers | |
Northern Analysis Running Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.