Learn More
Invitrogen™ NorthernMax™ Denaturing Gel Buffer (10X)
For preparing denaturing formaldehyde/MOPS agarose gels
Brand: Invitrogen™ AM8676
Code : 50
Additional Details : Weight : 0.36000kg
Description
- Gels made with this buffer are considerably less brittle than conventional formaldehyde gels, making handling of the gels much easier
- Formaldehyde is included
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- Ready to use and are certified RNase-free
- Comprehensive instructions are included
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 4°C. | |
10X | |
Northern Analysis Gel Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.