Learn More
Invitrogen™ NorthernMax™ Formaldehyde Load Dye
Ready-to-use solution
Brand: Invitrogen™ AM8552
Code : 50
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Description
- To use, add solution to RNA sample (3 parts solution: 1 part sample) and heat briefly:The samples are then ready to be loaded
- Ethidium bromide can be added to the samples if desired
- Compatible with formaldehyde-containing gels
- The same solution used in Invitrogen kits
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at -20°C. | |
Northern Analysis Loading Dye | |
Formaldehyde-Containing Gels | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.