Learn More
Invitrogen™ NorthernMax™-Gly Gel Prep/Running Buffer (10X)
For the preparation and running of agarose gels
Brand: Invitrogen™ AM8678
403.83 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10500kg
Description
- For use with RNA denatured by glyoxal treatment
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- These reagents are supplied ready to use and are certified RNase-free
- Comprehensive instructions are included
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 4°C. | |
10X | |
Northern Analysis Gel Prep/Running Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.