Learn More
Invitrogen™ NorthernMax™-Gly Sample Loading Dye
Used for RNA sample denaturation in a glyoxal gel protocol
Brand: Invitrogen™ AM8551
Code : 50
Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg
Description
- The volume ratio of solution to sample is lower than in many protocols, so sample precipitation prior to gel loading is usually not required
- Ethidium bromide is premixed into the solution
- Supplied in six tubes containing 1mL each
- Rigorously tested for nonspecific endonuclease activity, exonuclease activity, RNase activity, and for functionality
- Compatible with glyoxal-containing gels
- The same solution supplied in Invitrogen kits
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at -20°C. | |
Northern Analysis Sample Loading Dye | |
Room Temperature |
Ethidium bromide | |
Glyoxal-Containing Gels |
For Research Use Only. Not for use in diagnostic procedures.