Learn More
Invitrogen™ NorthernMax™ Low Stringency Wash Buffer
The equivalent of 2X SSC or SSPE, a low stringency wash buffer
Brand: Invitrogen™ AM8673
Code : 50
Additional Details : Weight : 0.10500kg
Description
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- Ready to use and are certified Rnase-free, tested functionally by northern analysis
- Comprehensive instructions are included
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
Wash Buffer | |
Northern Analysis Wash Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.