Learn More
Invitrogen™ NorthernMax™ One-Hour Transfer Buffer
Faster and more efficient than standard, overnight transfer buffer
Brand: Invitrogen™ AM8672
Code : 50
Additional Details : Weight : 0.10500kg
Description
- Transfer is complete in 1.5 to 2 hr
- Ready to use and are certified Rnase-free
- Transfer buffer is not compatible with nitrocellulose membrane
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- Comprehensive instructions are included
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
Northern Analysis Transfer Buffer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.