missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucAway™ Spin Columns

Product Code. 10279144
Click to view available options
:
Spin Columns
This item is not returnable. View return policy

Product Code. 10279144

Brand: Invitrogen™ AM10070

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For fast removal of unincorporated nucleotides and salts from probes after probe synthesis reactions

Invitrogen™ NucAway™ Spin Columns are for the fast removal of unincorporated nucleotides and salts from probes after probe synthesis reactions. 30 columns are provided.
  • Fast, efficient removal of unincorporated nucleotides and salts after probe synthesis
  • RNase- and DNase-free
  • Replaces more time- consuming purification methods To use, simply rehydrate the column, centrifuge to remove the interstitial fluid, add the probe to the top of the column, and centrifuge again to elute the probe.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Filtered Filtered
For Use With (Application) Removal of unincorporated nucleotides and salts after probe synthesis
Format Spin Column
Tip Style Universal Fit
Product Type NucAway™ Spin Columns
Content And Storage Store tubes at room temperature.
Shipping Condition Room Temperature
Disposable Single-use
Product Line Ambion, NucAway

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.