missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free

Product Code. 10722497
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10722497

Brand: Invitrogen™ AM9625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions.

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions which, when diluted to a 1X working concentration, contains 137 mM NaCl, 2.7 mM KCl, 8 mM Na2HPO4, and 2 mM KH2PO4. Each 10X PBS solution is ready to use upon dilution to the desired concentration. This molecular biology-grade PBS is certified RNase-free; it is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
pH 7.4
Physical Form Liquid
Product Line Ambion
Quantity 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.