missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PK Buffer for MagMAX™-96 DNA Multi-Sample Kit

This is the PK Buffer found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more PK Buffer than is provided in the kit. DNase-free.

Brand:  Invitrogen™ 4489111

17.40 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 15342506

  • £23.20 / 96 units

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: 2 tubes
This item is not returnable. View return policy

Description

Description

This is the PK Buffer found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more PK Buffer than is provided in the kit. DNase-free.

For use with: 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion Personal Genome Machine™ (PGM™) System, KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor, OpenArray™

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 bottle

Store at room temperature.
Magnetic Bead
Genomic DNA
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
96 Preps
DNA Multi-Sample Kit
DNase-Free
1 hr.
96-well Plate
Blood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
MagMAX™
96 Preps
Genomic DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.