missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Poly(A) Polymerase (cloned) 2 U/μL

Product Code. 10390495
Click to view available options
Quantity:
80 units, 2 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10390495

Brand: Invitrogen™ AM2030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Poly(A) Polymerase, 5X Poly(A) Polymerase Buffer, and 25mM MnCl2

Catalyzes the addition of adenosine to the 3' end of RNA in a sequence-independent fashion

  • The Invitrogen Poly(A) Polymerase enzyme can be used to append radiolabeled ATP to the 3' end of RNA molecules to generate labeled RNAs
  • One tube containing 80U (2U/μL), 5X Poly(A) Polymerase Buffer, and 25mM MnCl2 are provided
  • The enzyme can be used in conjunction with terminal transferase, reverse transcriptase, and thermostable polymerase to clone RNAs of unknown sequence
  • Unit Definition: One unit is the amount of enzyme required to incorporate 1nmol of AMP into acid-insoluble material in 10 minutes at 37°C

DNA & RNA Purification & Analysis, Nucleic Acid Labeling & Oligo Synthesis, PCR & Real-Time PCR, RNA Labeling, Reverse Transcription

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Poly(A) Polymerase, 5X Poly(A) Polymerase Buffer, and 25 mM MnCl2 should be stored at –20°C.
Shipping Condition Dry Ice
Polymerase Poly(A) RNA Polymerase
Product Line Ambion
Quantity 80 units, 2 μL
Concentration 5X
Source RNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.