Learn More
Invitrogen™ Poly(A) Tailing Kit
For the polyadenylation of in vitro transcribed RNA to enhance translation initiation efficiency.
Brand: Invitrogen™ AM1350
228.00 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Description
For the polyadenylation of in vitro transcribed RNA to enhance translation initiation efficiency. Sufficient reagents are included for 25 reactions.
- Adds a poly(A) tail of at least 150 nucleotides in length to the 3' termini of RNA
- Enhances translational efficiency in vivo
- Reaction can be adjusted to control for tail length
- Optimized for use with RNA transcripts synthesized using the mMESSAGE mMACHINE™ Kit
Specifications
E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Control DNA Template should be stored at -20°C. Nuclease-free Water may be stored at any temperature. |
|
Tailing Kit | |
Dry Ice | |
In Vitro Transcribed RNA (Polyadenylated) | |
RNA (In Vitro Transcribed) |
Kit | |
Ambion™ | |
miRNA analysis | |
25 Reactions |
For Research Use Only. Not for use in diagnostic procedures.